Thermo Scientific Chemicals

L-(+)-Ascorbic acid, Electrophoresis Grade, 99+%, Thermo Scientific Chemicals

Catalog number: J66291.18
50 g, Each
Thermo Scientific Chemicals

L-(+)-Ascorbic acid, Electrophoresis Grade, 99+%, Thermo Scientific Chemicals

Catalog number: J66291.18
50 g, Each
Quantity
Catalog number: J66291.18
also known as J66291-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
50-81-7
IUPAC Name
(5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2,5-dihydrofuran-2-one
Molecular Formula
C6H8O6
InChI Key
CIWBSHSKHKDKBQ-DOMZIZNONA-N
SMILES
OC[C@H](O)[C@H]1OC(=O)C(O)=C1O
Form
Powder or crystals or crystalline powder
Residue on ignition
≤0.1%
Optical Rotation
+20.5 to +21.5°
Assay from Supplier's CofA
≥99.0%
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text