DPDPE
Thermo Scientific Chemicals

DPDPE

For cellular and molecular biology applications
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mg
5 mg
Catalog number J66293.MCR
also known as J66293-M^
Price (USD)/ Each
64.70
-
Add to cart
Quantity:
1 mg
Request bulk or custom format
Price (USD)/ Each
64.70
Add to cart
DPDPE
Catalog numberJ66293.MCR
Price (USD)/ Each
64.70
-
Add to cart
Specifications
Purification MethodHPLC
Common Name[D-Pen2, 5] Enkephalin hydrate
FormatSolid
Molecular Weight (g/mol)645.79
Product TypeDPDPE
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C