Thermo Scientific Chemicals

Methyl 2,3-O-isopropylidene-beta-D-ribofuranoside, 98%, Thermo Scientific Chemicals

Catalog number: J66296.06
5 g, Each
Thermo Scientific Chemicals

Methyl 2,3-O-isopropylidene-beta-D-ribofuranoside, 98%, Thermo Scientific Chemicals

Catalog number: J66296.06
5 g, Each
Quantity
Catalog number: J66296.06
also known as J66296-06
Price (USD)
Quantity
-
Have Questions?

Description

Methyl 2,3-O-isopropylidene-beta-D-ribofuranoside is used for the manufacture of fine chemicals, and Industrial use resulting in manufacture of intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2,3-O-isopropylidene-beta-D-ribofuranoside is used for the manufacture of fine chemicals, and Industrial use resulting in manufacture of intermediates.

Solubility
Soluble in chloroform, dichloromethane, ethanol, ethyl acetate, and methanol

Notes
Light Sensitive & Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text