Netilmicin sulfate is used as a broad spectrum antibiotic active against most Gram-negative and some Gram-positive bacteria. It is a semisynthetic 1-N-ethyl derivative of sisomycin, an aminoglycoside antibiotic with action similar to gentamicin, but has less ear and kidney toxicity. It is used in aminoglycoside resistance studied
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Netilmicin sulfate is used as a broad spectrum antibiotic active against most Gram-negative and some Gram-positive bacteria. It is a semisynthetic 1-N-ethyl derivative of sisomycin, an aminoglycoside antibiotic with action similar to gentamicin, but has less ear and kidney toxicity. It is used in aminoglycoside resistance studied
Solubility
Soluble in water
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
RUO – Research Use Only
William A. Craig.; Sigurdur Gudmundsson.; Richard M. Reich . Netilmicin Sulfate: A Comparative Evaluation of Antimicrobial Activity, Pharmacokinetics, Adverse Reactions and Clinical Efficacy. Pharmacotherapy: The Journal of Human Pharmacology and Drug Therapy. 1983, 3, (6), 305-314.
Adams, E.; Puelings, D.; Rafiee, M.; Roets, E.; Hoogmartens, J. Determination of netilmicin sulfate by liquid chromatography with pulsed electrochemical detection. Journal of Chromatography A. 1998, 812, (1-2), 151-157.