Thermo Scientific Chemicals

D-Fructose-6-phosphate disodium salt, 95%

Catalog number: J66311.MC
100 mg, Each
Thermo Scientific Chemicals

D-Fructose-6-phosphate disodium salt, 95%

Catalog number: J66311.MC
100 mg, Each
Quantity
Catalog number: J66311.MC
also known as J66311-MC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
26177-86-6
Appearance (Color)
White
Assay from Suppliers CofA
≥94.0% (1H NMR)
Form
Powder or crystalline powder or crystals

Description

D-Fructose-6-phosphate disodium salt is a sugar intermediate of the glycolytic pathway that may be used to help identify, differentiate and characterize enzymes such as phosphofructokinase(s), pyrophosphate-dependent fructose-6-phosphate 1-phosphotransferase(s), D-fructose-6-phosphate aldolase(s), glutamine:fructose-6-phosphate amidotransferase(s) and glucosamine-6P synthase(s).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Fructose-6-phosphate disodium salt is a sugar intermediate of the glycolytic pathway that may be used to help identify, differentiate and characterize enzymes such as phosphofructokinase(s), pyrophosphate-dependent fructose-6-phosphate 1-phosphotransferase(s), D-fructose-6-phosphate aldolase(s), glutamine:fructose-6-phosphate amidotransferase(s) and glucosamine-6P synthase(s).

Solubility
Soluble in water (100 mg/ml).

Notes
Store at -20°C. Incompatible with heat and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text