Endothelin-3, Human
Thermo Scientific Chemicals

Endothelin-3, Human

For cellular and molecular biology applications
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 mg
1 mg
Catalog number J66312.EXD
also known as J66312-!D
Price (USD)/ Each
587.00
-
Add to cart
Quantity:
0.5 mg
Request bulk or custom format
Price (USD)/ Each
587.00
Add to cart
Endothelin-3, Human
Catalog numberJ66312.EXD
Price (USD)/ Each
587.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
SpeciesHuman
Purification MethodHPLC
FormatSolid
Molecular Weight (g/mol)2643.04
Product TypeEndothelin-3, human
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C