Thermo Scientific Chemicals

Icariin, 98%, Thermo Scientific Chemicals

Catalog number: J66324.MD
250 mg, Each
Thermo Scientific Chemicals

Icariin, 98%, Thermo Scientific Chemicals

Catalog number: J66324.MD
250 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J66324.MD
also known as J66324-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
489-32-7
IUPAC Name
5-hydroxy-2-(4-methoxyphenyl)-8-(3-methylbut-2-en-1-yl)-7-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-3-{[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}-4H-chromen-4-one
Molecular Formula
C33H40O15
InChI Key
TZJALUIVHRYQQB-XLRXWWTNSA-N
SMILES
COC1=CC=C(C=C1)C1=C(O[C@@H]2O[C@@H](C)[C@H](O)[C@@H](O)[C@H]2O)C(=O)C2=C(O)C=C(O[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)C(CC=C(C)C)=C2O1
Appearance (Color)
Pale yellow
Form
Powder
Assay from Supplier's CofA
≥97.5%
Water Content
≤5.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text