Thermo Scientific Chemicals

2,3-O-Isopropylidene-D-glyceraldehyde, 50% w/w in dichloromethane, Thermo Scientific Chemicals

Catalog number: J66337.06
5 g, Each
Thermo Scientific Chemicals

2,3-O-Isopropylidene-D-glyceraldehyde, 50% w/w in dichloromethane, Thermo Scientific Chemicals

Catalog number: J66337.06
5 g, Each
Quantity
Catalog number: J66337.06
also known as J66337-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15186-48-8
IUPAC Name
(4R)-2,2-dimethyl-1,3-dioxolane-4-carbaldehyde
Molecular Formula
C6H10O3
InChI Key
YSGPYVWACGYQDJ-YFKPBYRVSA-N
SMILES
CC1(C)OC[C@@H](O1)C=O
Appearance (Color)
Colorless to pale yellow
Form
Liquid
Assay (GC)
97.0% min.

Description

Reactant involved in organic reactions including, Henry condensation reactions, Synthesis of highly functionalized chiral aziridines, Olefination of aldehydes, C-H oxidations, Aldol-type condensations, Synthesis of hydrophilic pyrazine dyes for use as exogenous fluorescent tracer agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant involved in organic reactions including, Henry condensation reactions, Synthesis of highly functionalized chiral aziridines, Olefination of aldehydes, C-H oxidations, Aldol-type condensations, Synthesis of hydrophilic pyrazine dyes for use as exogenous fluorescent tracer agents.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text