Reactant involved in organic reactions including, Henry condensation reactions, Synthesis of highly functionalized chiral aziridines, Olefination of aldehydes, C-H oxidations, Aldol-type condensations, Synthesis of hydrophilic pyrazine dyes for use as exogenous fluorescent tracer agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Reactant involved in organic reactions including, Henry condensation reactions, Synthesis of highly functionalized chiral aziridines, Olefination of aldehydes, C-H oxidations, Aldol-type condensations, Synthesis of hydrophilic pyrazine dyes for use as exogenous fluorescent tracer agents.
Solubility
Difficult to mix.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Steinar Hagen.; Thorleif Anthonsen.; Lars Kilaas. Addition of diazomethane and sulfur ylides to the oxo-group in derivatives of ketoses and aldoses : I. Reactions of 2,3-O-isopropylidene-D-glyceraldehyde. Tetrahedron. 1979, 35, (21), 2583-2589.
Brigitte Hafele.; Volker Jager. A Short Synthesis of (S)-5-Hydroxy-2-penten-4-olide from D-Mannitol via 2,3-O-Isopropylidene-D-glyceraldehyde. Liebigs Annalen der Chemie. 1987, 1987, (1), 85-87.