Product Image
Thermo Scientific Chemicals

Oxyntomodulin

For cellular and molecular biology applications
Have Questions?
Quantity:
1 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66339.MCR
also known as J66339-M^
Price (USD)/ Each
427.00
-
Add to cart
Quantity:
1 mg
Request bulk or custom format
Price (USD)/ Each
427.00
Add to cart
Oxyntomodulin
Catalog numberJ66339.MCR
Price (USD)/ Each
427.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
SpeciesPorcine
Purification MethodHPLC
FormatSolid
Molecular Weight (g/mol)4421.90
Product TypeOxyntomodulin
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C