Cefmetazole sodium, 921μg/mg, Thermo Scientific Chemicals
Cefmetazole sodium, 921μg/mg, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cefmetazole sodium, 921μg/mg, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66348.03
also known as J66348-03
Price (USD)/ Each
100.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
100.00
Add to cart
Cefmetazole sodium, 921μg/mg, Thermo Scientific Chemicals
Catalog numberJ66348.03
Price (USD)/ Each
100.00
-
Add to cart
Specifications
Chemical Name or MaterialCefmetazole sodium
TypeCefmetazole Sodium
Name Note921μg/mg
Physical FormPowder
CAS56796-39-5
View more
Cefmetazole is used to study the effect of expression, binding, and inhibition of penicillin-binding proteins (with the exception of PBP2) on bacterial cell wall mucopeptide synthesis. It is also used to study protein mediated antibiotic transport.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Cefmetazole is used to study the effect of expression, binding, and inhibition of penicillin-binding proteins (with the exception of PBP2) on bacterial cell wall mucopeptide synthesis
  • It is also used to study protein mediated antibiotic tr
RUO – Research Use Only