Thermo Scientific Chemicals

D-Fructose-1,6-bisphosphate trisodium salt

Catalog number: J66349.03
1 g, Each
Thermo Scientific Chemicals

D-Fructose-1,6-bisphosphate trisodium salt

Catalog number: J66349.03
1 g, Each
Quantity
Catalog number: J66349.03
also known as J66349-03
Price (USD)
Quantity
-
Have Questions?

Description

D-Fructose-1,6-bisphosphate trisodium salt is used to reduce ischemia-reperfusion injury in rats.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Fructose-1,6-bisphosphate trisodium salt is used to reduce ischemia-reperfusion injury in rats.

Solubility
Soluble in water at 50mg/ml

Notes
Protect from heat. Incompatible with heat and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text