Thermo Scientific Chemicals

Xylenecyanol FF, Electrophoresis Reagent, dye content 70%, Thermo Scientific Chemicals

Catalog number: J66377.09
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Xylenecyanol FF, Electrophoresis Reagent, dye content 70%, Thermo Scientific Chemicals

Catalog number: J66377.09
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: J66377.09
also known as J66377-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2650-17-1
IUPAC Name
sodium 4-{[4-(ethylamino)-3-methylphenyl][(1Z,4Z)-4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl}-3-sulfobenzene-1-sulfonate
Molecular Formula
C25H27N2NaO6S2
InChI Key
VVLFAAMTGMGYBS-KRQUPCAFSA-M
SMILES
[Na+].CCNC1=C(C)C=C(C=C1)C(=C1/C=C\C(=N\CC)\C(C)=C1)\C1=C(C=C(C=C1)S([O-])(=O)=O)S(O)(=O)=O
Assay from Supplier's CofA
Dye Content: ≥70.0%
Assay from Supplier's CofA
Extinction Co-efficient (612-618 nm): ≥65000
Assay from Supplier's CofA
Spectrophotometric Assay (at 612-618 nm): Report
Comment
Suitable as a tracking dye for DNA sequencing
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text