Thermo Scientific Chemicals

Adrenomedullin (1-52), Human

Catalog number: J66379.MCR
1 mg, Each
Thermo Scientific Chemicals

Adrenomedullin (1-52), Human

Catalog number: J66379.MCR
1 mg, Each
Quantity
Catalog number: J66379.MCR
also known as J66379-M^
Price (USD)
Quantity
-
Have Questions?
Assay from Suppliers CofA
Peptide Purity: Report Value
Appearance (Color)
White
Form
Lyophilized Powder
Mass Spec
Exhibits correct MW

Description

Peptide elicits potent hypotensive effect, comparable to calcitonin gene-related peptide. Plays a key role in regulating endometrial angiogenesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text