Thermo Scientific Chemicals

Itraconazole, 98%, Thermo Scientific Chemicals

Catalog number: J66390.06
5 g, Each
Thermo Scientific Chemicals

Itraconazole, 98%, Thermo Scientific Chemicals

Catalog number: J66390.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J66390.06
also known as J66390-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
84625-61-6
IUPAC Name
1-(butan-2-yl)-4-{4-[4-(4-{[(2R,4S)-2-(2,4-dichlorophenyl)-2-[(1H-1,2,4-triazol-1-yl)methyl]-1,3-dioxolan-4-yl]methoxy}phenyl)piperazin-1-yl]phenyl}-4,5-dihydro-1H-1,2,4-triazol-5-one
Molecular Formula
C35H38Cl2N8O4
InChI Key
VHVPQPYKVGDNFY-ZPGVKDDISA-N
SMILES
CCC(C)N1N=CN(C1=O)C1=CC=C(C=C1)N1CCN(CC1)C1=CC=C(OC[C@H]2CO[C@@](CN3C=NC=N3)(O2)C2=CC=C(Cl)C=C2Cl)C=C1
Appearance (Color)
White to cream
Form
Powder
Assay (special)
ò 98.5% (USP method)
Proton NMR
Conforms to structure

Description

Itraconazole is a triazole antifungal agent. It is used to inhibit cytochrome P-450-dependent enzymes and ergosterol synthesis. It has been used against histoplasmosis, blastomycosis, cryptococcal meningitis, and aspergillosis. It?s different formulations are used to study Candida strains in murine invasive infections. It has been used to study proliferative changes of the forestomach mucosa in alloxan-induced diabetic rats..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Itraconazole is a triazole antifungal agent. It is used to inhibit cytochrome P-450-dependent enzymes and ergosterol synthesis. It has been used against histoplasmosis, blastomycosis, cryptococcal meningitis, and aspergillosis. It′s different formulations are used to study Candida strains in murine invasive infections. It has been used to study proliferative changes of the forestomach mucosa in alloxan-induced diabetic rats..

Solubility
Insoluble in water. Solube in chloroform at 50 mg/ml. Slightly soluble in ethanol or methanol

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text