Thermo Scientific Chemicals

Methicillin sodium salt, 90+%, Thermo Scientific Chemicals

Catalog number: J66414.03
1 g, Each
Thermo Scientific Chemicals

Methicillin sodium salt, 90+%, Thermo Scientific Chemicals

Catalog number: J66414.03
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J66414.03
also known as J66414-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
132-92-3
IUPAC Name
sodium (2S,5R,6R)-6-(2,6-dimethoxybenzamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular Formula
C17H19N2NaO6S
InChI Key
MGFZNWDWOKASQZ-UMLIZJHQSA-M
SMILES
[Na+].COC1=CC=CC(OC)=C1C(=O)N[C@H]1[C@H]2SC(C)(C)[C@@H](N2C1=O)C([O-])=O
Form
Powder
Water Content
≤10.0%
Appearance (Color)
White to off-white
Purity
≥90.0%

Description

Targets primarily the cell wall of Gram-positive organisms especially Staphylococcus aureus. Methicillin is used to study bacterium susceptibility, and to inhibit cell-wall synthesis. PBP's are enzymes which catalyze a pentaglycine crosslink between alanine and lysine residues providing additional strength to the cell wall. Without a pentaglycine crosslink, the integrity of the cell wall is severely compromised and ultimately leads to cell lysis and death. Methicillin Sodium Salt is inactivated by gastric acid so it is given to cells by other means in which gastric acid is not present in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Targets primarily the cell wall of Gram-positive organisms especially Staphylococcus aureus. Methicillin is used to study bacterium susceptibility, and to inhibit cell-wall synthesis. PBP′s are enzymes which catalyze a pentagl
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text