Thermo Scientific Chemicals

Piperacillin sodium salt, 835-1007^mg/mg, Thermo Scientific Chemicals

Catalog number: J66419.06
5 g, Each
Thermo Scientific Chemicals

Piperacillin sodium salt, 835-1007^mg/mg, Thermo Scientific Chemicals

Catalog number: J66419.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J66419.06
also known as J66419-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
59703-84-3
IUPAC Name
sodium (2S,5R,6R)-6-[(2S)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular Formula
C23H26N5NaO7S
InChI Key
WCMIIGXFCMNQDS-QMJSIVKPSA-M
SMILES
[Na+].CCN1CCN(C(=O)N[C@H](C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C([O-])=O)C2=CC=CC=C2)C(=O)C1=O
Form
Powder
Appearance (Color)
White to off-white
Assay from Supplier's CofA
863-1007 ug/mg

Description

A broad spectrum antibiotic targeting a wide range of susceptible gram positive and gram negative bacteria. Piperacillin sodium salt is an antibiotic. It is used in pharmacokinetic studies to optimize antimicrobial therapy and in hypersensitivity reactions. It is involved in the study of multidrug-resistant organisms. It acts as an antibiotic related to penicillin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in ethanol, water and methanol.

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use. Store in cool place. Incompatible with acids, heavy metal salts and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text