Thermo Scientific Chemicals

Decyl beta-D-glucopyranoside, Thermo Scientific Chemicals

Catalog number: J66423.03
1 g, Each
Thermo Scientific Chemicals

Decyl beta-D-glucopyranoside, Thermo Scientific Chemicals

Catalog number: J66423.03
1 g, Each
Quantity
Catalog number: J66423.03
also known as J66423-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
58846-77-8
IUPAC Name
(2R,3R,4S,5S,6R)-2-(decyloxy)-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Formula
C16H32O6
InChI Key
JDRSMPFHFNXQRB-IBEHDNSVSA-N
SMILES
CCCCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O
Appearance (Color)
White to off-white
Form
Powder or crystals or crystalline powder
Proton NMR
Conforms to structure
Purity
≥90.0%

Description

Non-ionic detergent used for solubilizing membrane proteins and in the development of sugar-based surfactant solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Non-ionic detergent used for solubilizing membrane proteins and in the development of sugar-based surfactant solutions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text