Thermo Scientific Chemicals

Minocycline hydrochloride, Thermo Scientific Chemicals

Catalog number: J66429.ME
500 mg, Each
Thermo Scientific Chemicals

Minocycline hydrochloride, Thermo Scientific Chemicals

Catalog number: J66429.ME
500 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J66429.ME
also known as J66429-ME
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13614-98-7
IUPAC Name
hydrogen (2Z,4S,4aS,5aR,12aS)-2-[amino(hydroxy)methylidene]-4,7-bis(dimethylamino)-10,11,12a-trihydroxy-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione chloride
Molecular Formula
C23H28ClN3O7
InChI Key
KDLQIOPKJDNQIM-WUURTAMISA-N
SMILES
[H+].[Cl-].CN(C)[C@H]1[C@@H]2C[C@@H]3CC4=C(C=CC(O)=C4C(O)=C3C(=O)[C@]2(O)C(=O)C(=C(N)O)C1=O)N(C)C
Form
Powder or crystals or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
Yellow
Assay from Suppliers CofA
≥97.5%

Description

A broad spectrum antibiotic commonly used against bacteria which cause upper respiratory tract infections

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A broad spectrum antibiotic commonly used against bacteria which cause upper respiratory tract infections
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text