N-Acetyl-Ser-Asp-Lys-Pro inhibits the entry of pluripotent hemopoietic stem cells into S-phase of the cell cycle and protects against Ara-C lethality in mice. It is a specific substrate for the N-terminal active site of angiotensin converting enzyme, which is responsible for its degradation in vivo.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Acetyl-Ser-Asp-Lys-Pro inhibits the entry of pluripotent hemopoietic stem cells into S-phase of the cell cycle and protects against Ara-C lethality in mice. It is a specific substrate for the N-terminal active site of angiotensin
RUO – Research Use Only
C S Anthony; G Masuyer; E D Sturrock; K R Acharya. Structure based drug design of angiotensin-I converting enzyme inhibitors. Current Medicinal Chemistry.2012, 19, (6), 845-855.
A Rousseau-Plasse; J Wdzieczak-Bakala; M Lenfant; E Ezan, R Genet; S Robinson; T Briscoe; J Melville; A Riches. Lisinopril, an angiotensin I-converting enzyme inhibitor, prevents entry of murine hematopoietic stem cells into the cell cycle after irradiation in vivo. Experimental Hematology.1998, 26, (11), 1074-1079.