Gly-Phe-Arg
Thermo Scientific Chemicals

Gly-Phe-Arg

 
Have Questions?
Quantity:
20 mg
Catalog number J66436.FPL
also known as J66436-F+
Price (USD)/ Each
71.30
-
Add to cart
Quantity:
20 mg
Request bulk or custom format
Price (USD)/ Each
71.30
Add to cart
Gly-Phe-Arg
Catalog numberJ66436.FPL
Price (USD)/ Each
71.30
-
Add to cart
Specifications
Purification MethodHPLC
FormatSolid
Molecular Weight (g/mol)378.40
Product TypeGly-Phe-Arg
Quantity20 mg
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C