Pipemidic acid, 98%, Thermo Scientific Chemicals
Pipemidic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pipemidic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66437.22
also known as J66437-22
Price (USD)/ Each
531.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
531.00
Add to cart
Pipemidic acid, 98%, Thermo Scientific Chemicals
Catalog numberJ66437.22
Price (USD)/ Each
531.00
-
Add to cart
Specifications
Chemical Name or MaterialPipemidic acid
TypePipemidic Acid
Physical FormPowder
CAS51940-44-4
Health Hazard 1H317-H334
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only