Thermo Scientific Chemicals
cAMP Dependent Protein Kinase Inhibitor (PKI 5-24)
Catalog number: J66438.MCR
1 mg, Each
Thermo Scientific Chemicals
cAMP Dependent Protein Kinase Inhibitor (PKI 5-24)
Catalog number: J66438.MCR
1 mg, Each
Quantity
Catalog number: J66438.MCR
also known as J66438-M^
Price (USD)
Quantity
-
Have Questions?
Description
cAMP Dependent Protein Kinase Inhibitor (PKI 5-24) acts as inhibitor of c-AMP-dependent protein kinase These proteins bind to the catalytic subunit of cyclic-AMP dependent protein kinase, displacing the regulatory subunit and inhibiting phosphorylating activity. The cyclic-AMP binding capacity of the regulatory subunit is enhanced.