Thermo Scientific Chemicals

2,3-O-Isopropylidene-D-ribonic acid-1,4-lactone, 97+%, Thermo Scientific Chemicals

Catalog number: J66442.06
5 g, Each
Thermo Scientific Chemicals

2,3-O-Isopropylidene-D-ribonic acid-1,4-lactone, 97+%, Thermo Scientific Chemicals

Catalog number: J66442.06
5 g, Each
Quantity
Catalog number: J66442.06
also known as J66442-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
30725-00-9
IUPAC Name
6-(hydroxymethyl)-2,2-dimethyl-tetrahydro-2H-furo[3,4-d][1,3]dioxol-4-one
Molecular Formula
C8H12O5
InChI Key
NHHKFJCWLPPNCN-UHFFFAOYNA-N
SMILES
CC1(C)OC2C(CO)OC(=O)C2O1
Assay (unspecified)
Specific rotation: -65? to -71?
Assay (HPLC)
>97.5%

Description

2,3-O-Isopropylidene-D-ribonic acid-1,4-lactone is used for the synthesis of C-nucleosides, prostanoids and GABA analogs via homochiral 2-piperidones. It is the starting material for the preparation of biochemical compounds including C-nucleosides, such as neplanocin A, and GABA analogs, via homochiral 2-piperidones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-O-Isopropylidene-D-ribonic acid-1,4-lactone is used for the synthesis of C-nucleosides, prostanoids and GABA analogs via homochiral 2-piperidones. It is the starting material for the preparation of biochemical compounds including C-nucleosides, such as neplanocin A, and GABA analogs, via homochiral 2-piperidones.

Solubility
Soluble in methanol at 50mg/ml, acetone, dichloromethane, ethyl Acetate.

Notes
Soluble in methanol at 50mg/ml
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text