Thermo Scientific Chemicals

Cefdinir, 95%, Thermo Scientific Chemicals

Catalog number: J66447.06
5 g, Each
Thermo Scientific Chemicals

Cefdinir, 95%, Thermo Scientific Chemicals

Catalog number: J66447.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J66447.06
also known as J66447-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
91832-40-5
IUPAC Name
(6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(N-hydroxyimino)acetamido]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
Molecular Formula
C14H13N5O5S2
InChI Key
RTXOFQZKPXMALH-GHXIOONMSA-N
SMILES
NC1=NC(=CS1)C(=NO)C(=O)N[C@H]1[C@H]2SCC(C=C)=C(N2C1=O)C(O)=O
Water Content (Karl Fischer Titration)
≤ 2.0%
Potency
960 to 1020 µg/ mg (on dry basis)
Appearance (Color)
White to light yellow
Identification (FTIR)
Conforms to structure
Optical Rotation
-61?C to -67?C

Description

A broad spectrum antibiotic targeting both Gram-positive and Gram-negative pathogens

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A broad spectrum antibiotic targeting both gram positive and gram negative pathogens
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text