Thermo Scientific Chemicals

Sphingosine-1-phosphate

Catalog number: J66459.MCR
1 mg, Each
Thermo Scientific Chemicals

Sphingosine-1-phosphate

Catalog number: J66459.MCR
1 mg, Each
Quantity
Catalog number: J66459.MCR
also known as J66459-M^
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
26993-30-6
IUPAC Name
{[(2S,3R,4E)-2-amino-3-hydroxyoctadec-4-en-1-yl]oxy}phosphonic acid
Molecular Formula
C18H38NO5P
InChI Key
DUYSYHSSBDVJSM-KRWOKUGFSA-N
SMILES
CCCCCCCCCCCCC\C=C\[C@@H](O)[C@@H](N)COP(O)(O)=O
Form
Powder or crystals or crystalline powder
Appearance (Color)
White to yellow
Infrared spectrum
Conforms
Assay from Supplier's CofA
≥97.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text