Thermo Scientific Chemicals

Ceftazidime pentahydrate, 95%, Thermo Scientific Chemicals

Catalog number: J66460.06
5 g, Each
Thermo Scientific Chemicals

Ceftazidime pentahydrate, 95%, Thermo Scientific Chemicals

Catalog number: J66460.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J66460.06
also known as J66460-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
78439-06-2
IUPAC Name
1-{[(6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-[(1-carboxy-1-methylethoxy)imino]acetamido]-2-carboxylato-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl}-1,2-dihydropyridin-2-ylium pentahydrate
Molecular Formula
C22H32N6O12S2
InChI Key
NMVPEQXCMGEDNH-TZVUEUGBSA-N
SMILES
O.O.O.O.O.CC(C)(ON=C(/C(=O)N[C@H]1[C@H]2SCC(CN3[CH+]C=CC=C3)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1)C(O)=O
Form
Powder or crystals or crystalline powder
Loss on Drying
13.0-15.0%
Assay from Supplier's CofA
≥94.0%
Appearance (Color)
White to off-white

Description

Ceftazidime pentahydrate is an antibacterial agent. It is used especially for Pseudomonas and other gram-negative infections in debilitated patients. It is used in the treatment of patients with infections caused by susceptible strains of organisms in the following diseases: lower respiratory tract infections,skin and skin structure infections, urinary tract infections, bacterial septicemia, bone and joint infections, gynecologic infections, intra abdominal infections (including peritonitis), and central nervous system infections (including meningitis).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Sparingly soluble in aqueous solution

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text