Thermo Scientific Chemicals

4-Nitrophenylphosphorylcholine, Thermo Scientific Chemicals

Catalog number: J66465.ME
500 mg, Each
Thermo Scientific Chemicals

4-Nitrophenylphosphorylcholine, Thermo Scientific Chemicals

Catalog number: J66465.ME
500 mg, Each
Quantity
Catalog number: J66465.ME
also known as J66465-ME
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
21064-69-7
IUPAC Name
4-nitrophenyl 2-(trimethylazaniumyl)ethyl phosphate
Molecular Formula
C11H17N2O6P
InChI Key
NAIXASFEPQPICN-UHFFFAOYSA-N
SMILES
C[N+](C)(C)CCOP([O-])(=O)OC1=CC=C(C=C1)[N+]([O-])=O
Appearance (Color)
White
Form
Solid
Assay from Suppliers CofA
≥98.0%
Proton NMR
Conforms to structure

Description

O-(4-Nitrophenylphosphoryl)choline is a useful probe in the assay of Phospholipase C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-(4-Nitrophenylphosphoryl)choline is a useful probe in the assay of Phospholipase C.

Solubility
Soluble in water

Notes
Moisture & Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text