Thermo Scientific Chemicals

D-Fructose-1,6-diphosphate dicalcium salt, 95%

Catalog number: J66477.14
25 g, Each
Thermo Scientific Chemicals

D-Fructose-1,6-diphosphate dicalcium salt, 95%

Catalog number: J66477.14
25 g, Each
Quantity
Catalog number: J66477.14
also known as J66477-14
Price (USD)
Quantity
-
Have Questions?

Description

D-Fructose-1,6-diphosphate dicalcium salt is essential for glycolysis to occur efficiently. It links to adenine nucleotides which regulate 6-phosphofructokinases (Pfks) that catalyze one of the rate limiting steps of glycolysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Fructose-1,6-diphosphate dicalcium salt is essential for glycolysis to occur efficiently. It links to adenine nucleotides which regulate 6-phosphofructokinases (Pfks) that catalyze one of the rate limiting steps of glycolysis.

Solubility
Soluble in water

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text