Thermo Scientific Chemicals

Cloxacillin sodium monohydrate, Thermo Scientific Chemicals

Catalog number: J66500.03
1 g, Each
Thermo Scientific Chemicals

Cloxacillin sodium monohydrate, Thermo Scientific Chemicals

Catalog number: J66500.03
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J66500.03
also known as J66500-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7081-44-9
IUPAC Name
sodium (2S,5R,6R)-6-[3-(2-chlorophenyl)-5-methyl-1,2-oxazole-4-amido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate hydrate
Molecular Formula
C19H19ClN3NaO6S
InChI Key
KCUWTKOTPIUBRI-VICXVTCVSA-M
SMILES
O.[Na+].CC1=C(C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C([O-])=O)C(=NO1)C1=CC=CC=C1Cl
Appearance (Color)
White to cream
Comment
Mass Spectrum: Consistent with structure
Assay from Supplier's CofA
≥98.0% (HPLC)
Form
Powder

Description

Targets primarily Gram-positive bacteria, especially --lactamase producing Staphylococci

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Targets primarily gram positive bacteria, especially _x001A_-lactamase producing Staphylococci
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text