Informational:Informational:This product is animal origin and can only be shipped within the United States
Thermo Scientific Chemicals

Cathepsin D, Human liver

Catalog number: J66506.EXF
25 μg, Each
Thermo Scientific Chemicals

Cathepsin D, Human liver

Catalog number: J66506.EXF
25 μg, Each
Quantity
Catalog number: J66506.EXF
also known as J66506-!F
Price (USD)
Have Questions?
Form
Solid
Assay (special)
Specific Activity: Report Value, post lyophilization
Purity
≥95.0%

Description

Cathepsin D, human liver is useful for inhibitor screening, kinetic studies, and cleavage of substrates in vitro or in tissue culture. It contains the mannose-6-phosphate lysosomal localization signal that targets the enzyme to the lysosomal compartment where it functions in the normal degradation of proteins. It is a useful prognostic indicator for breast cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text