Thermo Scientific Chemicals

Puerarin, 98%, Thermo Scientific Chemicals

Catalog number: J66528.09
10 g, Each
Thermo Scientific Chemicals

Puerarin, 98%, Thermo Scientific Chemicals

Catalog number: J66528.09
10 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J66528.09
also known as J66528-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3681-99-0
IUPAC Name
7-hydroxy-3-(4-hydroxyphenyl)-8-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-4H-chromen-4-one
Molecular Formula
C21H20O9
InChI Key
HKEAFJYKMMKDOR-VPRICQMDSA-N
SMILES
OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)C1=C2OC=C(C(=O)C2=CC=C1O)C1=CC=C(O)C=C1
Appearance (Color)
White to yellow
Assay from Supplier's CofA
≥97.5% (HPLC)
Form
Crystals or powder or crystalline powder
Proton NMR
Conforms to structure

Description

Thought to interfere with the synthesis of mycolic acids, an essential fatty acid component found in Mycobacterium cell walls

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Thought to interfere with the synthesis of mycolic acids, an essential fatty acid component found in Mycobacterium cell walls
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text