Thermo Scientific Chemicals

2,3:4,5-Di-O-isopropylidene-beta-D-fructopyranose, 98%

Catalog number: J66537.22
100 g, Each
Thermo Scientific Chemicals

2,3:4,5-Di-O-isopropylidene-beta-D-fructopyranose, 98%

Catalog number: J66537.22
100 g, Each
Quantity
Catalog number: J66537.22
also known as J66537-22
Price (USD)
Quantity
-
Have Questions?

Description

2,3:4,5-Di-O-isopropylidene-beta-D-fructopyranose is used as chiral auxiliaries in Michael and Aldol addition reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3:4,5-Di-O-isopropylidene-beta-D-fructopyranose is used as chiral auxiliaries in Michael and Aldol addition reactions.

Solubility
Soluble in chloroform, ethyl acetate, methanol.

Notes
Protect from heat. Incompatible with heat and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text