Cecropin A, Porcine
Thermo Scientific Chemicals

Cecropin A, Porcine

For cellular and molecular biology applications
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mg
0.5 mg
Catalog number J66546.MCR
also known as J66546-M^
Price (USD)/ Each
430.00
-
Add to cart
Quantity:
1 mg
Request bulk or custom format
Price (USD)/ Each
430.00
Add to cart
Cecropin A, Porcine
Catalog numberJ66546.MCR
Price (USD)/ Each
430.00
-
Add to cart
Specifications
SpeciesPorcine
Purification MethodHPLC
FormatLyophilized Powder
Molecular Weight (g/mol)4003.77
Product TypeCecropin A, porcine
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C