Myelin Oligodendrocyte Glycoprotein Peptide (35-55), Rat, Mouse
Myelin Oligodendrocyte Glycoprotein Peptide (35-55), Rat, Mouse
Thermo Scientific Chemicals

Myelin Oligodendrocyte Glycoprotein Peptide (35-55), Rat, Mouse

For cellular and molecular biology applications
Have Questions?
Quantity:
1 mg
Catalog number J66557.MCR
also known as J66557-M^
Price (USD)/ Each
373.00
-
Add to cart
Quantity:
1 mg
Request bulk or custom format
Price (USD)/ Each
373.00
Add to cart
Myelin Oligodendrocyte Glycoprotein Peptide (35-55), Rat, Mouse
Catalog numberJ66557.MCR
Price (USD)/ Each
373.00
-
Add to cart
Specifications
SpeciesMouse, Rat
Purification MethodHPLC
Common NameMOG Peptide (35-55)
FormatPowder
Molecular Weight (g/mol)2582.00
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C