Oxaliplatin
Thermo Scientific Chemicals

Oxaliplatin

 
Have Questions?
Quantity:
50 mg
Catalog number J66586.MF
also known as J66586-MF
Price (USD)/ Each
156.65
Online exclusive
174.00 
Save 17.35 (10%)
-
Add to cart
Quantity:
50 mg
Request bulk or custom format
Price (USD)/ Each
156.65
Online exclusive
174.00 
Save 17.35 (10%)
Add to cart
Oxaliplatin
Catalog numberJ66586.MF
Price (USD)/ Each
156.65
Online exclusive
174.00 
Save 17.35 (10%)
-
Add to cart
Specifications
Quantity50 mg
DescriptionWhite to off-white
Recommended StorageStore at -20°C
CAS61825-94-3
Molecular FormulaC8H14N2O4Pt, C8H14N2O4Pt
View more
Oxaliplatin is an anti-tumor agent with activity against colorectal cancer. It shows antitumor activity in cell lines with acquired cisplatin resistance. It is used to treat cancer of the gullet (oesophagus). It is used as adjuvant therapy in patients who have had surgery to remove the cancer. It is used to treat testicular cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C