Thermo Scientific Chemicals

Kainic acid monohydrate, 96%

Catalog number: J66590.MA
10 mg, Each
Thermo Scientific Chemicals

Kainic acid monohydrate, 96%

Catalog number: J66590.MA
10 mg, Each
Quantity
Catalog number: J66590.MA
also known as J66590-MA
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
58002-62-3
IUPAC Name
3-(carboxymethyl)-4-(prop-1-en-2-yl)pyrrolidine-2-carboxylic acid hydrate
Molecular Formula
C10H17NO5
InChI Key
FZNZRJRSYLQHLT-UHFFFAOYNA-N
SMILES
O.CC(=C)C1CNC(C1CC(O)=O)C(O)=O
Assay from Supplier's CofA
≥95.0%
Appearance (Color)
Cream
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text