[Met5, Lys6, Arg7] alpha-Neo-Endorphin (1-7), Thermo Scientific Chemicals
[Met<sup>5</sup>, Lys<sup>6</sup>, Arg<sup>7</sup>] alpha-Neo-Endorphin (1-7), Thermo Scientific Chemicals
Thermo Scientific Chemicals

[Met5, Lys6, Arg7] alpha-Neo-Endorphin (1-7), Thermo Scientific Chemicals

For cellular and molecular biology applications
Have Questions?
Quantity:
10 mg
Catalog number J66592.MA
also known as J66592-MA
Price (USD)/ Each
283.00
-
Add to cart
Quantity:
10 mg
Request bulk or custom format
Price (USD)/ Each
283.00
Add to cart
[Met5, Lys6, Arg7] alpha-Neo-Endorphin (1-7), Thermo Scientific Chemicals
Catalog numberJ66592.MA
Price (USD)/ Each
283.00
-
Add to cart
Specifications
Purification MethodHPLC
FormatSolid
Molecular Weight (g/mol)858.02
Product Type[Met5, Lys6, Arg7] alpha-Neo-Endorphin (1-7)
Quantity10 mg
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C