C-telopeptide
Thermo Scientific Chemicals

C-telopeptide

For cellular and molecular biology applications
Have Questions?
Quantity:
0.5 mg
Catalog number J66595.EXD
also known as J66595-!D
Price (USD)/ Each
110.00
-
Add to cart
Quantity:
0.5 mg
Request bulk or custom format
Price (USD)/ Each
110.00
Add to cart
C-telopeptide
Catalog numberJ66595.EXD
Price (USD)/ Each
110.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Purification MethodHPLC
Common NameC-terminal telopeptide
FormatSolid
Molecular Weight (g/mol)868.89
Product TypeC-telopeptide
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C