Allatostatin III
Thermo Scientific Chemicals

Allatostatin III

For cellular and molecular biology applications
Have Questions?
Quantity:
1 mg
Catalog number J66599.MCR
also known as J66599-M^
Price (USD)/ Each
69.70
-
Add to cart
Quantity:
1 mg
Request bulk or custom format
Price (USD)/ Each
69.70
Add to cart
Allatostatin III
Catalog numberJ66599.MCR
Price (USD)/ Each
69.70
-
Add to cart
Specifications
Purification MethodHPLC
FormatSolid
Molecular Weight (g/mol)899.00
Product TypeAllatostatin III
Quantity1 mg
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C