Thermo Scientific Chemicals

2-O-alpha-D-Glucopyranosyl-L-ascorbic acid, 98+%, Thermo Scientific Chemicals

Catalog number: J66601.06
5 g, Each
Thermo Scientific Chemicals

2-O-alpha-D-Glucopyranosyl-L-ascorbic acid, 98+%, Thermo Scientific Chemicals

Catalog number: J66601.06
5 g, Each
Quantity
Catalog number: J66601.06
also known as J66601-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
129499-78-1
IUPAC Name
(2R)-2-[(1S)-1,2-dihydroxyethyl]-5-hydroxy-4-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2,3-dihydrofuran-3-one
Molecular Formula
C12H18O11
InChI Key
YEERNPPBIMJBHH-DCWMUDTNSA-N
SMILES
OC[C@H](O)[C@H]1OC(O)=C(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)C1=O
Appearance (Color)
White to pale cream
Assay from Supplier's CofA
≥98+% (HPLC)
Form
Powder or crystalline powder
Proton NMR
Conforms to structure

Description

2-O-alpha-D-Glucopyranosyl-L-ascorbic acid reduce the free radicals that result from UV irradiation of the skin, and significantly reduce cell damage and photo-aging. It also promotes collagen synthesis. It can function as an antioxidant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-O-alpha-D-Glucopyranosyl-L-ascorbic acid reduce the free radicals that result from UV irradiation of the skin, and significantly reduce cell damage and photo-aging. It also promotes collagen synthesis. It can function as an antioxidant.

Solubility
Soluble in water. (879 g/L) at 25°C.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text