1x1 image pixel for data collection
Flucloxacillin sodium
Flucloxacillin sodium
Flucloxacillin sodium
Flucloxacillin sodium
Thermo Scientific Chemicals

Flucloxacillin sodium

Targets primarily the cell wall of susceptible gram-positive bacteria including Staphylococcus aureus. | CAS: 1847-24-1 | C19H16ClFN3NaO5S | 475.85 g/mol
Have Questions?
Catalog NumberQuantity
J66615.MC
also known as J66615-MC
100 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66615.MC
also known as J66615-MC
Price (USD)
105.00
Each
Add to cart
Quantity:
100 mg
Request bulk or custom format
Price (USD)
105.00
Each
Add to cart
Specifications
Boiling Point677°C
CAS1847-24-1
ChEBICHEBI:31615
Chemical Name or MaterialFlucloxacillin sodium
EINECS Number217-428-0
View more
Used in clinical in vitro tests known as antimicrobial susceptibility tests. It is used in treatment of pneumonia, bone infections, skin infections, wound infections and burns infections.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Flucloxacillin sodium is used to treat infections caused by susceptible Gram-positive bacteria. It is used in clinical in vitro tests known as antimicrobial susceptibility tests. It is used in treatment of pneumonia, bone infections, skin infections, wound infections and burns infections.

Solubility
Soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link