Thermo Scientific Chemicals

Apramycin sulfate, Thermo Scientific Chemicals

Catalog number: J66616.09
10 g, Each
Thermo Scientific Chemicals

Apramycin sulfate, Thermo Scientific Chemicals

Catalog number: J66616.09
10 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J66616.09
also known as J66616-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
65710-07-8
IUPAC Name
(2R,3R,4S,5S,6S)-2-{[(2R,3S,4R,4aR,6S,7R,8aS)-7-amino-6-{[(1R,2R,3S,4R,6S)-4,6-diamino-2,3-dihydroxycyclohexyl]oxy}-4-hydroxy-3-(methylamino)-octahydropyrano[3,2-b]pyran-2-yl]oxy}-5-amino-6-(hydroxymethyl)oxane-3,4-diol; sulfuric acid
Molecular Formula
C21H43N5O15S
InChI Key
WGLYHYWDYPSNPF-RQFIXDHTSA-N
SMILES
OS(O)(=O)=O.CN[C@H]1[C@@H](O)[C@H]2O[C@H](O[C@@H]3[C@@H](N)C[C@@H](N)[C@H](O)[C@H]3O)[C@H](N)C[C@@H]2O[C@@H]1O[C@H]1O[C@H](CO)[C@@H](N)[C@H](O)[C@H]1O
Loss on Drying
≤ 14%
Appearance (Color)
Pale yellow to yellow-brown powder
Potency
≥ 550 µg/mg (calculated on dry basis)
Form
Powder

Description

Apramycin sulfate binds to the deep groove of RNA and effectively inhibits ribosomal translocation prohibiting protein synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Apramycin is a 2-deoxysteptamine derivative and an aminoglycoside originally isolated from Streptomyces tenebrarius
• Apramycin sulfate is comprised of a bicyclic sugar moiety and a monosubstituted deoxystreptamine attached to a sulfate group

Application

• Apramycin sulfate exhibits antibiotic activity against gram-negative bacteria
• It is suitable for laboratory use against Escherichia coli, Klebsiella pneumoniae, and Pseudomonas aeruginosa
• It is also used for laboratory study of infectious disease

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text