Thermo Scientific Chemicals

Triton∣r X-100, Electrophoresis Reagent

Catalog number: J66624.AE
100 mL, Each
Thermo Scientific Chemicals

Triton∣r X-100, Electrophoresis Reagent

Catalog number: J66624.AE
100 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J66624.AE
also known as J66624-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9002-93-1
IUPAC Name
2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol
InChI Key
JYCQQPHGFMYQCF-UHFFFAOYSA-N
SMILES
CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1
Synonym
triton X polyoxyethylene (11) octyl phenyl ether polyoxyethylene (9) octylphenyl ether hydrol SW poletoxol peg-12 octyl phenyl ether triton X-102 ortho-gynol octoxynol octylphenoxypolyethoxyethanol
Comment
Color, Pt-Co: ≤60
Comment
Cloud Point: 63 to 69°C
Form
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text