Thermo Scientific Chemicals

Moxifloxacin hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: J66626.MC
100 mg, Each
Thermo Scientific Chemicals

Moxifloxacin hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: J66626.MC
100 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J66626.MC
also known as J66626-MC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
186826-86-8
IUPAC Name
hydrogen 1-cyclopropyl-6-fluoro-8-methoxy-7-{octahydro-1H-pyrrolo[3,4-b]pyridin-6-yl}-4-oxo-1,4-dihydroquinoline-3-carboxylic acid chloride
Molecular Formula
C21H25ClFN3O4
InChI Key
IDIIJJHBXUESQI-UHFFFAOYNA-N
SMILES
[H+].[Cl-].COC1=C2N(C=C(C(O)=O)C(=O)C2=CC(F)=C1N1CC2CCCNC2C1)C1CC1
Appearance (Color)
Yellow
Purity
≥96.0%
Form
Powder

Description

Moxifloxacin hydrochloride is a broad spectrum antibiotic commonly used against bacteria which cause sinus and lung infections. Mitoxantrone is a cytostatic anthracenedione that intercalates in DNA and increases the incidence of double-strand breaks by stabilizing the cleavable complex of topoisomerase II and DNA. Mitoxantrone also displays broad immunosuppressive activity inhibiting proliferation of all classes of lymphocytes and inducing apoptosis of antigen-presenting T cells. It used clinically as a chemotherapeutic agent against leukemias and solid tumors and as an immune system modulator in multiple sclerosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Moxifloxacin hydrochloride is a broad spectrum antibiotic commonly used against bacteria which cause sinus and lung infections. Mitoxantrone is a cytostatic anthracenedione that intercalates in DNA and increases the incidence of double-strand breaks by stabilizing the cleavable complex of topoisomerase II and DNA. Mitoxantrone also displays broad immunosuppressive activity inhibiting proliferation of all classes of lymphocytes and inducing apoptosis of antigen-presenting T cells. It used clinically as a chemotherapeutic agent against leukemias and solid tumors and as an immune system modulator in multiple sclerosis.

Solubility
Sparingly soluble in water. Soluble in DMSO

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text