Sarafloxacin hydrochloride hydrate, 98%, Thermo Scientific Chemicals
Sarafloxacin hydrochloride hydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sarafloxacin hydrochloride hydrate, 98%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
100 mg
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66639.MC
also known as J66639-MC
Price (USD)/ Each
28.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
33.30 
Save 4.65 (14%)
-
Add to cart
Quantity:
100 mg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
28.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
33.30 
Save 4.65 (14%)
Add to cart
Sarafloxacin hydrochloride hydrate, 98%, Thermo Scientific Chemicals
Catalog numberJ66639.MC
Price (USD)/ Each
28.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
33.30 
Save 4.65 (14%)
-
Add to cart
Chemical Identifiers
CAS1262886-26-9
SpecificationsSpecification SheetSpecification Sheet
Loss on Drying8.0% max.
Assay (unspecified)97.5% min.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only