Thermo Scientific Chemicals

Decyl beta-D-maltopyranoside, 97+%, Thermo Scientific Chemicals

Catalog number: J66644.06
5 g, Each
Thermo Scientific Chemicals

Decyl beta-D-maltopyranoside, 97+%, Thermo Scientific Chemicals

Catalog number: J66644.06
5 g, Each
Quantity
Catalog number: J66644.06
also known as J66644-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
82494-09-5
IUPAC Name
2-{[6-(decyloxy)-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Formula
C22H42O11
InChI Key
WOQQAWHSKSSAGF-UHFFFAOYNA-N
SMILES
CCCCCCCCCCOC1OC(CO)C(OC2OC(CO)C(O)C(O)C2O)C(O)C1O
Appearance (Color)
White to pale cream or pale yellow
Form
Crystals or powder or crystalline powder
Assay from Suppliers CofA
≥97.0%

Description

Decyl beta-D-maltopyranoside is used in a study to assess α-helical membrane protein crystallization. It has also been used in a study to investigate enantiomer separation of drugs by micellar electrokinetic chromatography. It aids in solubilizing the active cytochrome oxidase from mitochondria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Decyl beta-D-maltopyranoside is used in a study to assess α-helical membrane protein crystallization. It has also been used in a study to investigate enantiomer separation of drugs by micellar electrokinetic chromatography. It aids in solubilizing the active cytochrome oxidase from mitochondria.

Solubility
Soluble in methanol at 50mg/ml

Notes
Store at -20°C. Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text