Thermo Scientific Chemicals
Thidiazuron
Catalog number: J66663.MD
250mg, Plastic bottle, Each
Thermo Scientific Chemicals
Thidiazuron
Catalog number: J66663.MD
250mg, Plastic bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J66663.MD
Price (USD)
Quantity
-
Have Questions?
Description
Thidiazuron is a plant growth regulator used in micropropagations to supplement media such as Murashige and Skoog medium. TDZ promotes plant organogenesis and plant regeneration. Thidiazuron enables the production of virus-free stock plants, speeds up breeding, and provides new genotypes for the market.