Thermo Scientific Chemicals

Cholecystokinin, CCK Octapeptide (26-33)

Catalog number: J66669.MCR
1 mg, Each
Thermo Scientific Chemicals

Cholecystokinin, CCK Octapeptide (26-33)

Catalog number: J66669.MCR
1 mg, Each
Quantity
Catalog number: J66669.MCR
also known as J66669-M^
Price (USD)
Quantity
-
Have Questions?
Appearance (Color)
White
Assay from Suppliers CofA
Peptide Purity: ≥95.0%
Form
Lyophilized Powder
Mass Spec
Exhibits correct MW

Description

Cholecystokinin, CCK Octapeptide (26-33) is used in the biological studies in the preparation of opioid receptors agonistic and CCK receptor antagonistic activity. It plays a role in satiety.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text