Antide acetate, Thermo Scientific Chemicals
Antide acetate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Antide acetate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
1 mg
Catalog number J66692.LB0
also known as J66692-#0
Price (USD)/ Each
318.00
-
Add to cart
Quantity:
5 mg
Request bulk or custom format
Price (USD)/ Each
318.00
Add to cart
Antide acetate, Thermo Scientific Chemicals
Catalog numberJ66692.LB0
Price (USD)/ Each
318.00
-
Add to cart
Specifications
Chemical Name or MaterialAntide acetate
CAS112568-12-4
Health Hazard 1H360
Health Hazard 3P201-P202-P281-P308+P313-P501c
Storage Note 1-30° to -10°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only